missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), minus vitamin A
GREENER_CHOICE

Suitable for use in most neuronal cell applications

139.00 € - 1322.00 €

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Cell Type Neural Progenitor Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11500446
View Documents
Gibco™
12587010
10 mL
139.00 €
10mL
Please to purchase this item. Need a web account? Register with us today!
15440584
GREENER_CHOICE
View Documents
Gibco™
12587001
100 mL
1322.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Vitamin A is a customized version of the original B-27 Supplement without vitamin A (retinyl acetate). B-27 Supplement Minus Vitamin A is a serum-free supplement ideal for the cultivation of neural progenitor and stem cells, either as neurospheres in suspension or in adherent monolayer culture, without inducing differentiation.  B-27 Supplement Minus Vitamin A is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.

B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations and Neurobasal media

Specifications

Specifications

Store in freezer (-5 to -20°C) and protect from light.
Serum-free
Mammalian Cell Culture
Cell Culture Supplement
Sterile
Dry Ice
Neural Progenitor Cells
50 X
Liquid
Serum-Free
B-27™
Serum-Free
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

Product Identifier
  • 5-Methoxyindole

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Indol-5-yl methyl ether

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.