Learn More
Gibco™ B-27™ Supplement (50X), serum free
Used in the support of low or high density growth and short or long-term viability of hippocampal and CNS neurons. Gibco™ B27™ Supplement, Serum Free is suitable for use in most neuronal cell applications.
115.00 € - 1088.00 €
Specifications
Content And Storage | Store in freezer (-5 to -20°C) and protect from light. |
---|---|
Cell Type | Neural Cells |
Classification | Serum-free |
Culture Type | Mammalian Cell Culture |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11530536
|
Gibco™
17504044 |
10 mL |
115.00 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15360284
|
Gibco™
17504001 |
100 mL |
1088.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The most cited neuronal cell culture supplement, B-27 Supplement is an optimized serum-free supplement used to support the low- or high-density growth and short- or long-term viability of embryonic, post-natal, and adult hippocampal and other CNS neurons. B-27 Supplement is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.
- Provided as 50X liquid
- Used with Neurobasal™ Medium for neuronal cell culture
- Tested in growth assay utilizing rat (Sprague Dawley) embryonic hippocampal neurons
- Tested also for endotoxin at a 1X concentration
Specifications
Store in freezer (-5 to -20°C) and protect from light. | |
Serum-free | |
Liquid | |
Serum-Free | |
Endotoxin, Performance, Sterility, Mycoplasma |
Neural Cells | |
Mammalian Cell Culture | |
Cell Culture Supplement | |
Sterile |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.