missing translation for 'onlineSavingsMsg'
Learn More

Buchi Homogeniser B-400 with stainless steel knives 230V 50Hz 1900W Büchi

Brand:  Buchi 034220

Homogeniser B-400 with stainless steel knives 230V 50Hz 1900W Büchi

Code : 62

Additional Details : Weight : 28.75000kg

Product Code. 12821486

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers