Learn More
Brady™ B-427 Self-laminating Vinyl Thermal Transfer Labels
Printable, self-laminating vinyl labels for Thermal Transfer Printers.
Brand:  Brady™ 622025
 View more versions of this product
Product Code. 11579903
  • 297.00 € / 1 roll
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Matte
Thermal Transfer
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling
1
Square
0.102 mm
10 AWG to 4 AWG
38.1 mm
12.7 mm
UL Recognized
White, Transparent
Self-laminating Vinyl
Non-sterile
BBP™11,BBP™12, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
2500 Labels
-70°C to +70°C
41.3 mm
38.1 mm
38.1 mm