Learn More
Brady™ B-427 Self-laminating Vinyl Wire Marker Labels
Vinyl self-laminating labels for wire and electrical labeling.
Brand:  Brady™ 072013
 View more versions of this product
Product Code. 11563075
  • 718.00 € / 1 roll
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent acrylic adhesive
  • For smooth and textured surfaces
  • For matte finish
Specifications
Specifications
Not Autoclavable
Yellow, Transparent
Self-laminating Vinyl
Non-sterile
BBP™11, BBP™12, BradyPrinter i5100, i7100, PR Plus Series, and IP™ Series Printers, THT 76 mm Core Printers
3000 Labels
-70°C to +70°C
30.2 mm
25.4 mm
95.3 mm
Permanent Acrylic
Matte
Thermal Transfer
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
1
Rectangle
0.102 mm
1 AWG to 400 MCM
25.4 mm
25.4 mm