Learn More
Brady™ B-489A Polyester Labels for BBP33 and i3300 Printers
Polyester labels with permanent, rubber-based adhesive for use with B33 Series.
Brand:  Brady™ 142956
 View more versions of this product
Product Code. 15934550
  • 137.00 € / Pack of 500
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Permanent rubber adhesive
  • Abrasion resistant
  • For smooth, rough, or highly textured surfaces
Specifications
Specifications
Not Autoclavable
White
Polyester (B-483A)
Non-sterile
BBP™33, BradyPrinter i3300 and i5300 Industrial Label Printers
500 Labels
-40°C to +120°C
54 mm
50.8 mm
Permanent Rubber
Gloss
Thermal Transfer
Asset Tracking, Barcode Labeling, Equipment Labeling, Electrical Labeling, General Identification
1
Rectangle
0.1 mm
76.2 mm