missing translation for 'onlineSavingsMsg'
Learn More

Brady™ B-499 Nylon Cloth Label - B33 Series

The B-499 material has a protective polyamide coating, and permanent acrylic adhesive to stay adhered to surfaces. Stays in place even in autoclave, freezer, and liquid nitrogen environments.

Brand:  Brady™ 142995

Product Code. 16803423

  • 195.00 € / Pack of 1500
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Nylon cloth tape is ideal for use in cable and wire labelling, general-purpose marking in labs, equipment panels, and faceplates. Low-profile tape adheres to surfaces that are smooth, rough, or powder coated, as well as to irregular surfaces or surfaces with low surface energy. All-in-one cartridge includes ribbon and label supply that is UL approved and CSA accepted.

  • Label Properties: Permanent Adhesive Labels
  • Material: Nylon
  • Colour: White
  • For Use with Printer: BBP™33 Label Printer, BradyPrinter i3300
Specifications

Specifications

B33-125-499
White
BBP33, and BradyPrinter i3300 Label Printer
Acrylic
Polyamide-coated Nylon Cloth
22.8 x 12.7 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.