Learn More
Lonza™ b-G-Blocker
Glucan blocker to prevent false positive results in LAL assays.
Brand: Lonza™ N190
This item is not returnable.
View return policy
Description
- Glucan blocker to prevent false positive results during LAL assays.
- Some examples of glucan sources include yeast and cellulosic materials including hemodialysis filters.
- Product may be used with any of our LAL assays. Note: PYROGENT™ Gel Clot LAL products typically do not require ß-G-Blocker.
Specifications
LAL assays | |
Vials | |
Blocking glucan for LAL assays | |
5 x 5 mL |