missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ B-safe™ Polyethylene Biohazard Bag
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
135.00 € - 523.00 €
Specifications
Color | Yellow |
---|---|
Disposable | Single-use |
Material | Polyethylene |
Product Type | Biohazard Bag |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
12340609
|
Alpha Packaging™
12340609 |
255 mm |
135.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12300609
|
Alpha Packaging™
12300609 |
355 mm |
271.00 €
Pack of 50 |
Estimated Shipment: 11-03-2025
Log in to see stock. |
|||||
12320609
|
Alpha Packaging™
12320609 |
1015 mm |
523.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
Description
Manufactured from polyethylene. Alpha Packaging™ B-safe™ Polyethylene Biohazard Bags effectively contain biohazard waste.
- Comes in loose packs or benchtop dispenser boxes.
- For heavy-duty disposal
- Marked on both sides with biohazard symbol and warnings:
- "Biohazard, Danger of Infection"
- "Burn Without Opening"
- Warning in English, French, and German
- Polyethylene (500 gauge)
Specifications
Yellow | |
Single-use | |
Polyethylene | |
Biohazard Bag |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title