Learn More
Masterflex™ B/T™ Analog Drive
Masterflex™ B/T™ Analog Drive for use with high volume transfer and dispensing
Brand:  Masterflex™ 77111-37
Product Code. 13435539
  • 7680.00 € / Each
Estimated Shipment: 18-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Masterflex™ B/T™ drives are the best choice for high-flow bulk-transfer applications
  • Their durable construction ensures long service life with minimal maintenance
  • The sturdy materials of construction include stainless steel/carbon steel, a cast aluminum housing with an epoxy-powder coating, and Nylatron™ rollers for rugged dependability
  • All B/T™ drives include a new and improved Rapid-Load™ pump head for fast and easy tubing changes
  • The redesigned occlusion mechanism provides an improved tubing retention mechanism which holds the tubing firmly in place to further simplify the loading process
  • PerfectPosition™ tubing sizes provide improved performance in higher-pressure applications and provide longer tubing life
Specifications
Specifications
6.3 amps
(38.4 x 68.6 x 40 cm)
16 to 35 LPM
1 h/p
B/T™
Wash-Down Pump
Fixed-Speed
220 VAC
IP55-rated
3
B/T 87, or B/T 91
≤271 rpm