Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Nylon cloth labels for BBP33/i3300 printers.
Brand:  Brady™ 361679
 View more versions of this product
Product Code. 30041407
  • 243.00 € / Pack of 1000
Explore more special offers
Description
Description
For BBP33/i3300 printers.
Specifications
Specifications
B33-620-499
White
Nylon Cloth
Non-sterile
12.70 mm (W) x 25.40 mm (H)
12.70 mm
Permanent Acrylic
Matte
Labels
Rectangle
25.40 mm
12.70 mm