missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels
Nylon cloth labels for BBP33/i3300 printers.
Brand: Brady™ 361681
This item is not returnable.
View return policy
Description
For BBP33/i3300 printers.
Specifications
B33-621-499 | |
White | |
Nylon Cloth (B-499) | |
Non-sterile | |
25.40 mm (W) x 36.50 mm (H) | |
25.40 mm |
Permanent Acrylic | |
Matte | |
Labels | |
Rectangle | |
36.50 mm | |
25.40 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels > Brady™ Nylon Cloth Labels, Color; White, Dimensions; 25.40 mm (W) x 36.50 mm (H), Shape; Rectangle
Spot an opportunity for improvement?Share a Content Correction