Learn More
Brady™ B33 Series Polyamide Coated Nylon Cloth Labels with Vial Top
Nylon cloth labels for BBP33/i3300 printers.
Brand:  Brady™ 143011
Product Code. 30041433
  • 242.00 € / Pack of 1500
Explore more special offers
Description
Description
For BBP33/i3300 printers.
Specifications
Specifications
UL Recognized,CSA Accepted;Halogen Free (DIN VDE 0472/part 815) Approvals are for White material only
Permanent Acrylic
Matte
Labels
Laboratory labelling
1
Rectangle
-40°C to +90°C
0.166 mm
0.5 in.
0.9 in.
0.5 in.
Nylon Cloth Labels for BBP33/i3300 Printers
White
Nylon Cloth
Non-sterile
BBP33, i3300,i5300
1500 Stickers
-40°F to +194°F
0.0065 in.
22.86 mm (W) x 12.70 mm (H) x 12.70 mm (Dia)
12.70 mm
22.86 mm
12.70 mm