missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bac-to-Bac™ C-His TOPO™ Expression System
Bac-to-Bac™ C-His TOPO™ Expression System
Brand: Invitrogen™ A11100
2560.20 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Bac-to-Bac™ Baculovirus Expression is an efficient method for producing baculovirus for high-level protein expression in insect cells. It relies on generation of recombinant virus by site-specific transposition in E. coli rather than homologous recombination in insect cells. New: pFastBacTOPO™ vectors!Reliable and fast protein expression
The Bac-to-Bac™ Baculovirus Expression System is faster and easier than traditional baculovirus expression, and it maintains high levels of protein expression. Bac-to-Bac™ relies on generation of recombinant baculovirus by site-specific transposition in E. coli rather than homologous recombination in insect cells. The Bac-to-Bac™ Baculvirus Expression System is highly regarded in academic Literature. Approximately 100 citations every year since 2000!
Traditional baculovirus systems require purification and amplification of an initial low-titer viral supernatant. This requires a time-consuming plaque assay. Bac-to-Bac™'s pFastBac™ vector, however, recombines with the parent bacmid in DH10Bac™ E. coli competent cells to form an expression bacmid. Transfect the bacmid into insect cells for fast production of a high titer of pure recombinant baculovirus particles in the very first transfection. You'll save weeks of precious time. Collect a pure P2 baculovirus stock on Day 10 without the necessity of tedious plaques assay (See Figure 1)
High expression, easy screening
pFastBac™ uses the strong polyhedrin promoter to generate high levels of expression in a variety of insect cell lines such as Sf9, Sf21, and High Five™ cells.
New additions:
Bac-to-Bac™'s pFastBac vectors are now available as pFastBacTOPO™ vectors too. pFastBacTOPO™ vectors are Blunt™ TOPO™ vectors and are supplied with Mach1™-T1R E. coli for easy cloning and pFastBac⁄GOI propagation. The new vectors enable you to:
The new vectors combine the TOPO™ cloning technology with the highly regarded Bac-to-Bac™ baculovirus expression features to enable easy cloning and high-level protein expression.
The pFastBacTOPO™ vectors are offered as cloning and expression kits.
Specifications
Expression System | |
His-Tag (6x) | |
Bac-to-Bac C-His TOPO Expression Kit: • Vector kit for 20 TOPO cloning reactions, store at -20°C –pFastBac/CT-TOPO vector containing the C-terminal TEV cleavage sit and His-tag –pFastBac 1-Gus control plasmid (control expression vector) –Other kit reagents: 10X PCR, dNTP mix, Salt solution, sterile water, control PCR template, polyhedrin forward primer, SV40 pA reverse primer • One Shot Mach1-T1R Chemically Competent E. coli for 20 reactions (20 x 50 μL), store at -80°C • MAX Efficiency DH10Bac Competent E. coli for 20 reactions (4 kits with 5 x 0.1 mL each), store at -80°C • ExpiFectamine Sf Transfection Reagent (1 mL), store at 4°C, do not freeze |
|
Liquid | |
Baculovirus | |
Polyhedrin |
Blunt TOPO™ | |
20 Reactions | |
Ampicillin (AmpR), Gentamicin (GmR) | |
Cell-Based Expression | |
Bac-to-Bac™, TOPO™ |
For Research Use Only. Not for use in diagnostic procedures.