missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Background Plate, 384-well

Used to maintain your real-time PCR instrument with 384-well block

Brand:  Applied Biosystems™ 4481310

Product Code. 17146138

  • 139.00 € / Each
Estimated Shipment: 07-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 384-Well Background Plate is used to maintain your real-time PCR instrument with 384-well block.
  • Background calibration should be run at least every six months.
  • Please see your instrument user guide for full instructions on use.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at –20°C ± 5°C.

Background plate
384
1 x 384 Plate
ViiA™ 7 Dx System, QuantStudio™ Dx
1 x 384-well plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.