Learn More
Testo™ 610 Compact Humidity/Temperature Meter with Backlit Display
Use this thermohygrometer for a variety of applications. Testo™ 610 Compact Humidity/Temperature measures temperature and humidity in indoor areas. In addition, the compact instrument can also calculate the dew point and the wet bulb temperature.
Brand:  Testo™ 0560 0610
Product Code. 10365053
  • 295.00 € / Each
Explore more special offers
Description
Description
  • Easy temperature and humidity measurement
  • Calculation of dew point and wet bulb temperature
  • Illuminated display
  • Capacitive humidity sensor with long-term stability
Specifications
Specifications
0 to 100%
NTC: ±0.5°C; Humidity: ±2.5 %RH (5 to 95 %RH)
±0.5°C
119 x 46 x 25 mm (incl. protective cap)
Humidity: 0.1 %RH
-10°C to +50°C
0.1°C
90 g
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products