Learn More
Molecular Probes™ Bacteria Counting Kit, for flow cytometry
Descripción
The Bacteria Counting Kit accurately enumerates bacteria by flow cytometry. The kit includes the SYTO™ BC dye, which is also available separately (Cat No. S34855), a high-affinity nucleic acid stain that easily penetrates both gram-negative and gram-positive bacteria, producing an exceptionally bright green-fluorescent signal. The kit also includes a calibrated suspension of polystyrene microspheres. Signals from both the stained bacteria and the beads are easily detected in the green fluorescence channel and can be distinguished on a plot of forward scatter versus fluorescence. The density of the bacteria in the sample can be determined from the ratio of bacterial signals to microsphere signals in the cytogram.
Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.
Especificaciones
Especificaciones
| Product Type | Bacteria Counting Kit |
| Detection Method | Fluorescence |
| Quantity | 1 kit |
| Sample Type | Bacteria |
| Diameter (Metric) Bead | 6 μm |
| Content And Storage | Contains 1 vial of SYTO™ BC bacterial stain (100 μL, 1000x in DMSO), 1 vial microsphere standard (1 mL, 6 μm). Store in refrigerator (2–8°C) and protect from light. |
| For Use With (Application) | Flow Cytometry |
| For Use With (Equipment) | Flow Cytometer |
| Includes | BC Bacterial Stain, Microsphere Standard |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.