Learn More
Novacyt - Microgen Bioproducts Bacterial ID System Reagent/Consumable
Additional/supplemental reagent/consumable for biochemical identification system.
Brand: Novacyt - Microgen Bioproducts MID61E
This item is not returnable.
View return policy
Description
- Replacement/supplemental reagent/consumable.
- From a range of simple, reliable, bacterial identification systems for food, environmental, and industrial laboratories.
- These tests will arm professionals with the necessary information to maximize the effectiveness of appropriate patient management.
- The range is expanding quickly and will encompass the widest range of microbiological testing solutions.
Specifications
TDA Reagent | |
Biochemical Identification | |
Biochemical Identification Kits |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Supplemental information
- TDA REAGENT
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
Supplemental information
- MIXTURE LIST-Contains : Iron (III)Chloride,Hydrochloric acid