Learn More
Thomil™ Bacteromil Antiseptic Hand Gel
Thomil™ Bacteromil is a specially-formulated rinse-free friction antiseptic hand gel for healthy skin that is virucidal, bactericidal and levuricidal, and also efficacy-tested against coronavirus.
POA - 58.97 €
Specifications
Color | Light Blue |
---|---|
Certifications/Compliance (Europe) | UNE-EN 1500 (Antiseptic by friction); UNE-EN 13624 (Levuricide); UNE-EN 14476 (Virucide) |
Composition | Denatured Ethanol (70%), Triclosan (0.20%), Benzalkonium Chloride (0.16%) |
pH Range | Neutral |
Solution Type | Ethanolic |
Alert:
For external use only. Use biocides safely. Always read the label and product information before use. Do not use on people sensitive to any product components, or for extended periods of time. Do not mix with other products. Use with caution in children under two years. Highly flammable liquid and vapour. Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16876033
![]() |
Thomil™
HAHP061 |
2 Pk. | |||||||
16886033
![]() |
Thomil™
HAHP062 |
8 Bottles |
POA
Pack of 8 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16896033
![]() |
Thomil™
HAHP069 |
12 Bottles |
POA
Pack of 12 |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- A biocidal hand gel for complete, safe disinfection of healthy skin
- Synergistic mixture of alcohols and surfactants formulated to WHO recommendations with a neutral pH
- Complies with UNE-EN 1500 (Antiseptic by Friction), UNE-EN 13624 (Levuricide) and UNE-EN 14476 (Virucide) standards
- Quick drying with a smooth, non-sticky finish
- Does not require rinsing or subsequent drying
- Ideal for frequent use anywhere, especially where soap and water are not readily available
Specifications
Light Blue | |
Denatured Ethanol (70%), Triclosan (0.20%), Benzalkonium Chloride (0.16%) | |
Ethanolic |
UNE-EN 1500 (Antiseptic by friction); UNE-EN 13624 (Levuricide); UNE-EN 14476 (Virucide) | |
Neutral |
Safety and Handling
- X2 4L No Rinse Antiseptic Gel
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : ethanol, quarternaryammonium compounds,ethandiol, cyclohexane
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.