missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Casitone
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
10 kg
missing translation for 'unitSize'
10kg
500g
Beschreibung
- An animal origin (AO) pancreatic enzymatic digest of casein similar to Difco Casein Digest
- This supplement provides a nutritional profile based on casein
- The specific enzymatic digest performed for this product results in a particularly high content of amino acids and peptides of varying sizes, making it a more nutritious hydrolysate
- It has been successfully used for the production of Escherichia coli-based recombinant proteins
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Spezifikation
Spezifikation
| Certifications/Compliance | ISO/DIS, 11014 |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Form | Powder |
| Product Type | Dehydrated Culture Media |
| Format | Bottle |
| For Use With (Application) | Production of Escherichia coli-based recombinant proteins |
| Product Line | Bacto |
| Quantity | 500 g |
| Source | Animal origin, casein |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur