Learn More
Gibco™ Bacto™ Proteose Peptone No. 3
Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media.
Brand:  Gibco™ 212230
 View more versions of this product
Product Code. 16299771
  • 7270.00 € / 10kg
Explore more special offers
Description
Description
  • Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein
  • Studies show that no single peptone is the most suitable nitrogen source for every microbiological application
  • These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
  • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
  • Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LUO, MUO
Mammalian, Insect, Microbial
Bacterial Media
Enzymatic digest of Protein
10 kg
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Powder
Pail
Bacto™
Animal Origin