missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Bacto™ Tryptose

A mixed enzymatic hydrolysate with distinctive nutritional properties.

4750.00 €

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Product Line Bacto™
Source Animal Origin
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16289751
View Documents
Gibco™
211709
10 kg
4750.00 €
10kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
Animal Origin
Powder
Bacto™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.