Learn More
Interscience™ BagOpen 400 for DiluFlow
BagOpen 400 for dilutor.
Brand:  Interscience™ 505016
Product Code. 17545245
  • 697.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of durable stainless steel
Specifications
Specifications
Bag Stand
400 mL
DiluFlow, DiluFlow Pro, DiluFlow Elite
Not Autoclavable
Standing Up Bags
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Interscience™ BagOpen 400 for DiluFlow > BagOpen 400 for DiluFlow; Volume (Metric): 400 mL