missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Baird-Parker Agar (ISO) Base (Dehydrated)
Isolate, enumerate, and identify Staphylococcus aureus in food samples with selective Baird-Parker Agar (ISO) Base.
361.00 € - 2190.00 €
Specifications
Content And Storage | 10°C to 30°C |
---|---|
Description | Baird-Parker Agar (ISO) Base (Dehydrated) |
Form | Powder |
Product Type | Agar |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12731680
|
Thermo Scientific™
CM1127T |
5 kg |
2190.00 €
5kg |
Estimated Shipment: 12-02-2025
Log in to see stock. |
|||||
12721680
|
Thermo Scientific™
CM1127B |
500 g |
361.00 €
500g |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||||
Description
Isolate, enumerate, and identify coagulase positive Staphylococcus aureus in food samples with selective Thermo Scientific™ Oxoid™ Baird Parker Agar (ISO) Base (Dehydrated) while adhering to ISO 6888-1:1999.
Baird Parker Agar contains carefully balanced selective inhibitory agents, glycine, lithium, and tellurite, to suppress the growth of most of the bacteria present in foods without inhibiting coagulase positive staphylococci. The presence of egg yolk emulsion and sodium pyruvate aid the recovery of damaged cells for presumptive identification of Staphylococcus aureus in food specimens.Baird-Parker Agar base is widely recommended by national and international bodies for the isolation of Staphylococcus aureus1.
- Easy interpretation of test results as Staphylococcus aureus forms gray–black shiny colonies with surrounding clear zones.
- Adheres to ISO 6888-1:1999 standards.
Egg yolk emulsion makes the medium cream/yellow and opaque. Coagulase-positive staphylococci reduce tellurite to form black/grey shiny colonies which, after 48 hours, produce clear zones around the colonies caused by the proteolysis of egg yolk. On further incubation, most strains of coagulase-positive staphylococci form opaque haloes around the colonies.
Some strains of Staphylococcus saprophyticus may also produce both clear zones and opaque haloes but can be distinguished by the longer incubation required5.
Colonies typical of Staphylococcus aureus , but without an egg yolk reaction, should also be tested for coagulase production6. Egg yolk reaction negative Staphylococcus aureus may occur in some food, especially cheese.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
10°C to 30°C | |
Baird-Parker Agar (ISO) Base (Dehydrated) | |
Powder | |
Agar |
Safety and Handling
missing translation for 'shelfLife' : 3 Years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title