Learn More
R&D Systems™ BAK BH3 Synthetic Peptide GQVGRQLAIIGDDINR Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Brand:  R&D Systems™ 881-BA-100
Product Code. 18118550
  • 169.00 € / 100µg
Explore more special offers
Description
Description
The BAK BH3 Synthetic Peptide GQVGRQLAIIGDDINR Protein has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
578
BAK BH3 Domain, aa 72-87
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant
1.7 kDa
100 μg
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.