Learn More
R&D Systems™ BAK L to A BH3 Synthetic Peptide GQVGRQAAIIGDDINR
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity
Brand:  R&D Systems™ 879-BK-100
125.26 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18177210
  • 159.00 € / 100µg
Explore more special offers
Description
Description
The BAK L to A BH3 Synthetic Peptide GQVGRQAAIIGDDINR has been validated for the following applications: Inhibition Activity.
Specifications
Specifications
578
BAK L78A BH3 Domain, aa 72-87
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Recombinant
1.7 kDa