missing translation for 'onlineSavingsMsg'
Learn More

J.T. Baker BAKERBOND spe™ Silica Gel (SiOH) Disposable Extraction Columns, J.T. Baker™

245.00 € - 1690.00 €

Specifications

Column Phase Silica gel
Description Disposable extraction column
Format Granules
Particle Size 40 μm
Sorbent Silica
View More Specs

Products 4
Product Code Brand Column Capacity (Metric) Bed Weight Quantity Price Quantity & Availability  
Product Code Brand Column Capacity (Metric) Bed Weight Quantity Price Quantity & Availability  
10381511
View Documents Promotion Details
J.T. Baker
7086-03
3 mL 500 mg Pack of 50
289.00 €
Pack of 50
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
10474911
View Documents Promotion Details
J.T. Baker
7086-06
6 mL 500 mg Pack of 30
245.00 €
Pack of 30
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
10781272
View Documents Promotion Details
J.T. Baker
7086-07
6 mL 1000 mg Pack of 30
290.00 €
Pack of 30
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
15547654
View Documents Promotion Details
J.T. Baker
7086-26
6 mL 500 mg Pack of 250
1690.00 €
Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
 
Description

Description

Specifications

Specifications

Silica gel
Granules
Silica
White
Disposable extraction column
40 μm
60 angstroms
Videos
SDS
Documents

Documents

Product Certifications