Learn More
Cole-Parmer™ Balance Draft Shield
Clear acrylic prevents drafts from affecting weighings
Brand:  Cole-Parmer™ 01019-06
 View more versions of this product
Product Code. 15300655
  • 289.00 € / Each
Explore more special offers
Description
Description
Clear acrylic prevents drafts from affecting weighings
  • No assembly required
  • Mouse hole in back panel for power cords
  • Raised edges on top of enclosure allow secure placement of frequently used weighing accessories
  • Minimizes air movement around your balance for faster, more stable readings
Specifications
Specifications
Clear
19.75 x 18 x 18.5 in.
457 mm
Large