missing translation for 'onlineSavingsMsg'
Learn More

Balance Mix OLM04.1, SPEX CertiPrep™

Product Code. 15256326
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Acetophenone98-86-22000μg/mL
Atrazine1912-24-92000μg/mL
Benzaldehyde100-52-72000μg/mL
Biphenyl92-52-42000μg/mL
e-Caprolactam105-60-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 98-86-2
Packaging 2mL amber prescored glass ampule
Quantity 1 mL
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C8H8O
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC=C1
IUPAC Name 1-phenylethanone
Molecular Weight (g/mol) 120.151
PubChem CID 7410
ChEBI CHEBI:27632
Methods For US EPA CLP Methods
Solvent or Matrix Acetone
Chemical Name or Material Semi-Volatile Balance Mix, CLP Series
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'