Learn More
Thermo Scientific™ Balance Plate for Pierce™ 96-Well Filter Spin Plates
Balance Plate for Pierce™ 96-Well Filter Spin Plates
Brand: Thermo Scientific™ 45205
Code : A0
Additional Details : Weight : 0.10000kg
Description
These Thermo Scientific Plate Accessories are collection plates and centrifuge balance plates for affinity purification and desalting resin products that use Thermo Scientific Pierce 96-Well Filter Spin Plates.Related Products
Collection Plates (Polypropylene) for Pierce™ 96-Well Filter Spin Plates
Specifications
Balance Plate | |
Yes | |
Pierce™ |
96-well Plate | |
96-Well Plate |
For Research Use Only. Not for use in diagnostic procedures.