Learn More
Lenz Laborglasintrumente Ball Members Inch Range
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01213520
 View more versions of this product
Product Code. 10490123
  • 134.00 € / Pack of 10
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ball members
  • Inch range
  • Acc. to DIN 12264
  • Made of DURAN™ tubing
Specifications
Specifications
S 35
Duran Glass