missing translation for 'onlineSavingsMsg'
Learn More

Uvex Bamboo TwinFlex™ D xg Cut Protection Gloves
GREENER_CHOICE

The Bamboo TwinFlex™ technology combines the best of both worlds in one fine 18-gauge cut protection glove (Cut Level D).

201.00 €

Specifications

Certifications/Compliance (Europe) EN 388:2016 (4 X 4 1 D), EN 407 (X 1 X X X X)
Construction Seamless Knit
Coating Material Aqua-Polymer
Color Green
Hand Ambidextrous
View More Specs

Products 7
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17532905
GREENER_CHOICE
View Documents
Uvex
6009006
6
201.00 €
10 pairs
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
17542905
GREENER_CHOICE
View Documents
Uvex
6009007
7
201.00 €
10 pairs
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
17552905
GREENER_CHOICE
View Documents
Uvex
6009008
8
201.00 €
10 pairs
In Stock
to see stock.
Edge
Add to basket
 
17562905
GREENER_CHOICE
View Documents
Uvex
6009009
9
201.00 €
10 pairs
In Stock
to see stock.
Edge
Add to basket
 
17572905
GREENER_CHOICE
View Documents
Uvex
6009010
10
201.00 €
10 pairs
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
17582905
GREENER_CHOICE
View Documents
Uvex
6009011
11
201.00 €
10 pairs
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
17592905
GREENER_CHOICE
View Documents
Uvex
6009012
12
201.00 €
10 pairs
Estimated Shipment: 10-12-2024
to see stock.
Add to basket
 
Description

Description

Bamboo TwinFlex™ D xg: Sustainability, comfort and sensitivity combined with high cut protection (CutD)

The patented Bamboo TwinFlex™ technology combines the best of both worlds in one fine 18-gauge cut protection glove (Cut Level D). On the inside of the glove, wearers experience noticeable comfort for their skin thanks to the bamboo-viscose. The smooth fit and high moisture absorption with a rapid action provides long-lasting comfort and high wearer acceptance. With a flexible fit thanks to the bamboo-viscose, the glove adapts precisely to fit the shape of the hand within 3 to 5 minutes of wearing. High-performance fibers offer a high level of protection against abrasion and cut hazards. The additional protection zone in the area between the thumb and index finger provides double the abrasion resistance (200%) compared to the inside of the glove to increase its service life/durability. Thanks to the Xtra-Grip aqua-polymer foam coating, the Bamboo TwinFlex™ D xg offers excellent grip in dry and slightly damp/oily conditions. In addition, the Bamboo TwinFlex D xg features a total of 45% sustainable materials: recycled polyamide and bamboo-viscose, which is based on renewable raw materials.

General Features:

  • Excellent sensitivity in cut protection level D
  • High level of wearer comfort thanks to bamboo fibre
  • 45% sustainable materials: recycled polyamide and bamboo viscose, which is based on renewable raw materials
  • High efficiency due to long service life, all thanks to protection + premium Xtra-Grip coating
  • CO2-neutral production in Germany, free from solvents

Protective Features

  • Certified to EN 388:2016 (4 X 4 1 D) and to EN 407 (X 1 X X X X)
  • Dermatologically tested for skin compatibility, free from allergenic accelerators
  • Particularly light, thin, and flexible with high cut protection (4X41D according to EN 388:2016)
  • No need to remove the glove thanks to touchscreen capability protection zone with smooth material surface ensures trouble-free workflow in accordance with EN 407, the model is suitable for contact warmth up to +100 °C

Comfort Features

  • Patented Bamboo TwinFlex™ yarn technology
  • Silky soft feel against the skin
  • Fast absorption and high retention of moisture
  • Adaptive fit: Adapts precisely to fit the shape of any hand within 3 to 5 minutes of wearing

Areas of application

  • Installation of sharp-edged or unburred parts
  • Glass and metalworking industry, automotive industry
  • Precision work
  • Quality control
  • Areas with use of touchscreens food processing
Specifications

Specifications

EN 388:2016 (4 X 4 1 D), EN 407 (X 1 X X X X)
Aqua-Polymer
Ambidextrous
Bamboo-viscose, Elastane, HPPE, Polyamide, Steel
EN 388:4 X 4 1 D
Knit
Seamless Knit
Green
Yes
Cut-resistant Gloves
Unisex
ISO 9001; ISO 14001; OEKO-TEX™ Standard 100 , proDerm, uvex pure standard
Videos
SDS
Documents

Documents

Product Certifications