Learn More
Banoxantrone-d12 dihydrochloride, MedChemExpress
MedChemExpress Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.

446.85 € - 1366.20 €
Specifications
Chemical Name or Material | Banoxantrone-d12 dihydrochloride |
---|---|
CAS | 1562066-98-1 |
Color | Blue |
Purity Grade Notes | Research |
Molecular Formula | C22H18D12Cl2N4O6 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16418437
![]() |
Medchem Express
HY-13562AS |
5 mg |
1366.20 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16401359
![]() |
Medchem Express
HY-13562AS |
1 mg |
446.85 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Banoxantrone-d12 (AQ4N-d12) dihydrochloride is the deuterium labeled Banoxantrone dihydrochloride. Banoxantrone is a novel bioreductive agent that can be reduced to a stable, DNA-affinic compound AQ4, which is a potent topoisomerase II inhibitor.Specifications
Banoxantrone-d12 dihydrochloride | |
Blue | |
C22H18D12Cl2N4O6 | |
Cancer-programmed cell death | |
O=N(C([2H])([2H])[2H])(C([2H])([2H])[2H])CCNC1=CC=C(NCCN(C([2H])([2H])[2H])(C([2H])([2H])[2H])=O)C(C(C2=C(O)C=CC(O)=C23)=O)=C1C3=O.Cl.Cl | |
529.48 | |
Research |
1562066-98-1 | |
Research | |
AQ4N-d12 dihydrochloride | |
Solid | |
529.48 | |
98.02% |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.