missing translation for 'onlineSavingsMsg'
Learn More

Barban, SPEX CertiPrep™

Product Code. 16340218
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Barban101-27-91000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 101-27-9, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Color White
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C11H9Cl2NO2
InChI Key MCOQHIWZJUDQIC-UHFFFAOYSA-N
SMILES C1=CC(=CC(=C1)Cl)NC(=O)OCC#CCCl
IUPAC Name 4-chlorobut-2-ynyl N-(3-chlorophenyl)carbamate
Molecular Weight (g/mol) 258.098
PubChem CID 7551
ChEBI CHEBI:33421
Methods For US EPA Method 8270
Analyte or Component Names Barban
Solvent or Matrix Methanol
pH 8.7 to 9.3
Physical Form Powder/Solid
Chemical Name or Material Barban
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'