Learn More
JULABO™ Barbed Fittings
Barbed Fittings are fluid tubing adapters to convert male barbed fittings of various Internal Diameters (ID) to male G thread fittings with the same ID.
Brand: JULABO™ 8890152
This item is not returnable.
View return policy
Description
- Fitting styles include:
- 3/4 in. ID male to G 3/4 in. male
- 1 in. ID male to G 1 in. male
- 1 1/2 in. ID male to G 1 1/2 in. male
Specifications
1.5 in. ID Male to G 1.5 in. Male | |
Fitting Adapter, Fluid Tubing | |
2 Pk. |