Learn More
Corning™ Bar-coded Cryogenic Vials
For storing biological and compound samples in labeled vials at cryogenic temperatures. Corning™ Bar-coded Cryogenic Vials provides a validated unique bar-code that will not fade or fall off.
Brand: Corning™ 8670
This item is not returnable.
View return policy
Description
- Manufactured from polypropylene
- Leak proof seal for integrity of storage
- Autoclavable vials and caps, can be re-sterilized
- Unique 1D, 2D bar-codes and human readable characters
- Features star shaped cavity in cap and is compatible with decapping equipment
- Locking base stabilizes tubes for automated handling
- Graduations and writing patch for manual use also
- Each tube is checked for clarity and readability of 2D code
Specifications
Orange | |
Sterile | |
Cell biology, cell banking, compound cryopreservation | |
50 | |
Round, Self-standing | |
Polyethylene |
Natural | |
2 mL | |
500 | |
Internal | |
2D |
Product Content Correction
Product Title
Corning™ Bar-coded Cryogenic Vials > Internal; 1D and 2D
Spot an opportunity for improvement?Share a Content Correction