Learn More
Emma Safety Footwear Bari (Frontier 118) Safety Shoes
Made with the same care and attention to detail associated with high end luxury shoes. Brazilian 2-tone full grain leather combined with nubuck adds to the stylish look for a shoe that fits every occasion.
Brand:  Emma Safety Footwear MM11809046
 View more versions of this product
Product Code. 15948201
  • 231.00 € / 1 pair
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Thermoplastic Polyurethane Outsole
  • ESD certified
  • Chrome-free lining
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Blue
Hydro-Tec™ Sanitized Silver & Wellness leather
46
Laces
Bari (Frontier 118)
Leather
Steel