missing translation for 'onlineSavingsMsg'
Learn More
Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals Product Code.: 10712973

Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals

Product Code. 10712973
100 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 10712973

Brand: Thermo Scientific Acros 317861000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-39-3
Molecular Formula Ba
Molecular Weight (g/mol) 137.33
MDL Number MFCD00134031
InChI Key DSAJWYNOEDNPEQ-UHFFFAOYSA-N
Synonym bario, baryum, bario spanish, baryum french, barium, soluble compounds, unii-24gp945v5t, and soluble compounds, barium, elemental, and compounds, atomShow MoreShow Less
PubChem CID 5355457
ChEBI CHEBI:32595
IUPAC Name barium
SMILES [Ba]

Specifications

Color Gray
Melting Point 725.0°C
Boiling Point 1640.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula Ba
Quantity 100 g
Merck Index 15, 960
Formula Weight 137.34
Percent Purity ≥99%
Packaging Plastic bottle
Form Rod
Chemical Name or Material Barium, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAD4672792 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog Number
Lot NumberAC4650205 Certificate TypeCertificate of Analysis Date06/03/2025 Catalog Number
Lot NumberA0462799 Certificate TypeCertificate of Analysis Date17/05/2024 Catalog Number317865000,317861000
Lot NumberA0459926 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number317865000,317861000
Lot NumberA0451011 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number317865000,317861000
compliance-icons compliance-icons
Product Identifier
  • Barium in Paraffin oil
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Aspiration toxicity Category 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : White mineral oil, Barium

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Barium, 99+%, rods, ∽2cm diameter, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.