missing translation for 'onlineSavingsMsg'
Learn More

Barium acetate, ACS, 99.0-102.0%, Thermo Scientific Chemicals

Product Code. 11366598
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium acetate is used as a chemical reagent, textile mordant and lubricant. It is used to prepare other acetates and barium carbonate. It acts as a mordant for printing textile fabrics. It is utilized for drying paints and varnishes. It serves as a catalyst in organic synthesis. It plays an important role in the separation of glycosaminoglycans. Further, it acts as a cationization agent for sensitive multiple reaction monitoring of plasma free fatty acid profiling in a fish oil human intervention. In addition, it is used in the process of titrating sulfates of quinine and quinidine in nonaqueous conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 543-80-6
Molecular Formula C4H6BaO4
Molecular Weight (g/mol) 255.42
MDL Number MFCD00012447
InChI Key ITHZDDVSAWDQPZ-UHFFFAOYSA-L
Synonym barium acetate, barium diacetate, acetic acid, barium salt, octan barnaty, barium di acetate, caswell no. 068a, octan barnaty czech, unii-fba31yj60r, ccris 7240, barium 2+ diacetate
PubChem CID 10980
IUPAC Name barium(2+);diacetate
SMILES [Ba++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline
Quantity 250 g
UN Number UN1564
Merck Index 14,965
Solubility Information Soluble in water. Insoluble in alcohol.
Formula Weight 255.43
Percent Purity 99.0 to 102.0%
Grade ACS
Odor Odorless
Density 2.47 g/mL
Chemical Name or Material Barium acetate
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'