missing translation for 'onlineSavingsMsg'
Learn More

Barium bromate, 97%, Thermo Scientific Chemicals

Product Code. 11444473
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13967-90-3
Molecular Formula BaBr2O6
Molecular Weight (g/mol) 393.129
MDL Number MFCD00014177
InChI Key VEASZGAADGZARC-UHFFFAOYSA-L
Synonym barium bromate, bromic acid, barium salt, unii-5qk4064y0r, barium 2+ dibromate, acmc-1bumj, ba.2bro3, bromic acid, barium salt 2:1, barium bromate un2719 oxidizer
PubChem CID 61706
IUPAC Name barium(2+);dibromate
SMILES [O-]Br(=O)=O.[O-]Br(=O)=O.[Ba+2]

Specifications

Quantity 100 g
UN Number UN2719
Formula Weight 393.16
Percent Purity 97%
Odor Odorless
Density 3.99
Chemical Name or Material Barium bromate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'