missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide, 99%, pure, anhydrous, Thermo Scientific Chemicals

Product Code. 15239826
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For ultra trace analysis

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 10553-31-8
Molecular Formula BaBr2
Molecular Weight (g/mol) 297.16

Spécification

Melting Point 850.0°C
Quantity 250 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula BaBr2
Merck Index 15,964
Solubility Information Solubility in water: 104.1g/100mL (20°C). Other solubilities: soluble in methanol, almost insoluble in ethanol, ethyl acetate,, acetone, dioxane
Formula Weight 297.16
Percent Purity 99%
Grade Pure
Chemical Name or Material Barium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'