missing translation for 'onlineSavingsMsg'
Learn More

Barium bromide, anhydrous, 99%, Thermo Scientific Chemicals

Product Code. 11397208
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Barium bromide, anhydrous is used as the chemical intermediate, and the spice cosmetics chemical additive. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10553-31-8
MDL Number MFCD00003444

Specifications

Melting Point 850°C
Color White
Physical Form Powder
Quantity 250 g
UN Number UN1564
Merck Index 14,968
Solubility Information Soluble in water
Formula Weight 297.14
Percent Purity 99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.781 g/mL
Chemical Name or Material Barium bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'