missing translation for 'onlineSavingsMsg'
Learn More

Barium calcium copper oxide (2-1-2), 99.9% (metals basis), Thermo Scientific™

52.00 € - 212.00 €

Specifications

Chemical Name or Material Barium calcium copper oxide (2-1-2)
CAS 127922-05-8
Physical Form Powder
Assay Percent Notes (metals basis)
MDL Number MFCD00211652
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393568
View Documents Promotion Details
Thermo Scientific Alfa Aesar
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383568
View Documents Promotion Details
Thermo Scientific Alfa Aesar
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Barium calcium copper oxide (2-1-2)
Powder
MFCD00211652
Hydrolyzes with water.
127922-05-8
(metals basis)
UN1564
99.9%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c

H302+H332

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications