missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10412221
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point 1450.0°C
Melting Point 811.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99% min. (Complexometry)
Linear Formula BaCO3
Merck Index 15, 965
Solubility Information Solubility in water: 0.002g/100mL (20°C)
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Specific Gravity 4.43
Density 4.4300g/mL
Chemical Name or Material Barium carbonate, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'