missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, ACS, 99.0-101.0%, Thermo Scientific Chemicals

84.32 € - 244.22 €

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11363288
View Documents
Thermo Scientific Alfa Aesar
036236.36
500 g
84.32 €
500g
Please to purchase this item. Need a web account? Register with us today!
11373288
View Documents
Thermo Scientific Alfa Aesar
036236.A1
1 kg
134.67 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11353288
View Documents
Thermo Scientific Alfa Aesar
036236.D3
2 x 1 kg
244.22 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

513-77-9
197.34
AYJRCSIUFZENHW-UHFFFAOYSA-L
10563
CBaO3
MFCD00003448
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
[Ba++].[O-]C([O-])=O

Specifications

(decomposition)
513-77-9
Powder
CBaO3
barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
AYJRCSIUFZENHW-UHFFFAOYSA-L
barium(2+) carbonate
10563
ACS
White powder
Barium carbonate, For analysis ACS
811°C
White
99.0 to 101.0%
MFCD00003448
Soluble in most acids,with the exception of sulfuric acid. It is insoluble in water and ethanol.
[Ba++].[O-]C([O-])=O
197.34
197.35
Odorless
4.43 g/cm3
Videos
Safety and Handling

Safety and Handling

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 208-167-3

missing translation for 'rtecsNumber' : CQ8600000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.