missing translation for 'onlineSavingsMsg'
Learn More

Barium Carbonate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10121470
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
IUPAC Name barium(2+) carbonate
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point 1450°C
Melting Point 811°C
Color White
Physical Form Powder/Solid
Quantity 100 g
Formula Weight 197.35g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Barium carbonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'