missing translation for 'onlineSavingsMsg'
Learn More

Barium carbonate, tech., Thermo Scientific Chemicals

Product Code. 11355797
Click to view available options
Quantity:
100 g
500 g
2 kg
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers

Used in the manufacture of glass, brick, clay products, barium chemicals, barium ferrites, and production of photographic papers.Barium carbonate has been used as brine purification chemical in the chloro-alkali industry and as an ingredient in rat poisons and enamels. Barium carbonate is used in photographic paper manufacturing. The chemical is used to produce the barium sulphate which gives the photographic paper its flat white appearance. Practiced in the manufacture of barium ferrites and also in the manufacture of television picture tubes. Barium carbonate is an additive in oil-well drilling muds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-77-9
Molecular Formula CBaO3
Molecular Weight (g/mol) 197.34
MDL Number MFCD00003448
InChI Key AYJRCSIUFZENHW-UHFFFAOYSA-L
Synonym barium carbonate, carbonic acid, barium salt 1:1, barium carbonate 1:1, barium monocarbonate, carbonic acid, barium salt, pigment white 10, caswell no. 069, witherite, ci pigment white 10, bw-p
PubChem CID 10563
SMILES [Ba++].[O-]C([O-])=O

Specifications

Boiling Point (decomposition)
Melting Point 811°C
Physical Form Powder
Quantity 100 g
Merck Index 14,969
Solubility Information Almost insoluble in water; Soluble in NH4Cl solution,dilute HCl,HNO3,acetic acid; Insoluble in most alkalies;Soluble in most acids,with the exception of sulfuric acid; It is insoluble in water and ethanol.
IUPAC Name barium(2+) carbonate
Formula Weight 197.35
Grade Technical
Odor Odorless
Density 4.43
Chemical Name or Material Barium carbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'