missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride concentrate, For 1L standard solution, 0.05 M BaCl2 (0.1N), Honeywell Fluka™

Product Code. 15635190
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
This item is not returnable. View return policy
This item is not returnable. View return policy

for 1L standard solution, 0.05 M BaCl2 (0.1N)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10361-37-2
Molecular Formula BaCl2
Molecular Weight (g/mol) 208.23
MDL Number MFCD00003445
InChI Key WDIHJSXYQDMJHN-UHFFFAOYSA-L
Synonym barium chloride, barium dichloride, barium chloride solution, ccris 2286, barium chloride, anhydrous, barium chloride, ultra dry, bariumchlorid, dichlorobarium, dsstox_cid_24508
PubChem CID 25204
ChEBI CHEBI:63317
IUPAC Name barium(2+);dichloride
SMILES [Cl-].[Cl-].[Ba++]

Specifications

Quantity 1 Pc.
UN Number NONH for all modes of transport
Formula Weight 208.23g/mol
Chemical Name or Material Barium chloride concentrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'