missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium chloride concentrate, For 1L standard solution, 0.05 M BaCl2 (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
CAS Min % | ≥70.0000%, ≥10.0000% |
CAS Max % | <90.0000%, <20.0000% |
Physical Form | Concentrate |
Quantity | 6 x 1 Ea. |
Linear Formula | BaCl2 |
Formula Weight | 208.23 |
Packaging | Ampoule |
Density | 1.000 g/cm3 |
Chemical Name or Material | Barium chloride concentrate, For 1L standard solution, 0.05 M BaCl2 (0.1N) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'