Learn More
Barium chloride dihydrate, 99+%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10326-27-9 |
---|---|
Molecular Formula | BaCl2H4O2 |
Molecular Weight (g/mol) | 244.26 |
MDL Number | MFCD00149154 |
InChI Key | PWHCIQQGOQTFAE-UHFFFAOYSA-L |
Synonym | barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d |
PubChem CID | 5284346 |
ChEBI | CHEBI:86153 |
SMILES | O.O.[Cl-].[Cl-].[Ba++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11489020
|
Thermo Scientific Alfa Aesar
A12905.30 |
250 g |
N/A
|
N/A | |||||
11469020
|
Thermo Scientific Alfa Aesar
A12905.0B |
1000 g |
N/A
|
N/A | |||||
11479020
|
Thermo Scientific Alfa Aesar
A12905.0I |
5000 g |
N/A
|
N/A | |||||
Description
Barium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBarium chloride dihydrate is mainly used in the purification of brine solution in caustic chlorine plants, as a reagent in the test for sulfate ion in chemical lab, and as a cleansing agent in the production of other barium chemicals and lubricating oil additives. It is used in the production of molecular sieves, pigments, and paper coatings, and serves as a precursor to fabricate the doped barium zirconate.
Solubility
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate.
Notes
Stable and hygroscopic in nature.
Chemical Identifiers
10326-27-9 | |
244.26 | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
5284346 | |
O.O.[Cl-].[Cl-].[Ba++] |
BaCl2H4O2 | |
MFCD00149154 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
CHEBI:86153 |
Specifications
113°C (in 2H2O) | |
BaCl2H4O2 | |
UN1564 | |
barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d | |
PWHCIQQGOQTFAE-UHFFFAOYSA-L | |
barium(2+) dihydrate dichloride | |
5284346 | |
244.28 (208.25 Anhydrous) | |
Odorless | |
Barium chloride dihydrate |
10326-27-9 | |
MFCD00149154 | |
14,971 | |
Soluble in water and metanol. Insoluble in ethanol and ethyl acetate. | |
O.O.[Cl-].[Cl-].[Ba++] | |
244.26 | |
CHEBI:86153 | |
≥99% | |
3.097 |
Safety and Handling
GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
P261-P264b-P270-P271-P280i-P301+P310-P304+P340-P305+P351+P338-P312-P330-P501c
H301-H319-H332
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: BARIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 233-788-1
missing translation for 'rtecsNumber' : CQ8751000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only